accessibility menu, dialog, popup

UserName

NH2ClN

CAS RN 38487-86-4

IUPAC Name: 2-amino-4-chlorobenzonitrile
Synonyms: 2-amino-4-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: UZHALXIAWJOLLR-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)