missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzonitrile 98.0+%, TCI America™

Catalog No. A24835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24835G 5 g
A248325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24835G Supplier TCI America Supplier No. A24835G

Chemical Identifiers

CAS 38487-86-4
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD00035927
InChI Key UZHALXIAWJOLLR-UHFFFAOYSA-N
Synonym benzonitrile, 2-amino-4-chloro, 5-chloro-2-cyanoaniline, 4-chloro-2-aminobenzonitrile, 4-chloroanthranilonitrile, 2-amino-4-chlorobenzenecarbonitrile, pubchem20756, 2-amino-4-chloroberizonitrile, bidd:gt0102, 2-amino-4-chlorobenzonitrile, 2-amino-4-chloro-benzonitrile
PubChem CID 170059
IUPAC Name 2-amino-4-chlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)N)C#N

Specifications

Melting Point 163°C
Color Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 152.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Amino-4-chlorobenzonitrile