missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chlorobenzenethiol 98.0+%, TCI America™
$46.60 - $190.13
Chemical Identifiers
| CAS | 1004-00-8 |
|---|---|
| Molecular Formula | C6H6ClNS |
| Molecular Weight (g/mol) | 159.631 |
| MDL Number | MFCD00792528 |
| InChI Key | NGIRMPARLVGMPX-UHFFFAOYSA-N |
| Synonym | 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427 |
| PubChem CID | 12000 |
| IUPAC Name | 2-amino-4-chlorobenzenethiol |
| SMILES | C1=CC(=C(C=C1Cl)N)S |
Chemical Identifiers
| 1004-00-8 | |
| 159.631 | |
| NGIRMPARLVGMPX-UHFFFAOYSA-N | |
| 12000 | |
| C1=CC(=C(C=C1Cl)N)S |
| C6H6ClNS | |
| MFCD00792528 | |
| 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427 | |
| 2-amino-4-chlorobenzenethiol |
Specifications
| 1004-00-8 | |
| C6H6ClNS | |
| 5 g | |
| 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427 | |
| C1=CC(=C(C=C1Cl)N)S | |
| 159.631 | |
| 159.63 | |
| Crystalline Powder |
| 58°C | |
| MFCD00792528 | |
| 3259 | |
| NGIRMPARLVGMPX-UHFFFAOYSA-N | |
| 2-amino-4-chlorobenzenethiol | |
| 12000 | |
| ≥98.0% (GC,T) | |
| 2-Amino-4-chlorobenzenethiol |
Safety and Handling
TSCA : No