missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-nitrophenol 98.0+%, TCI America™

Catalog No. A224425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A224425G 25 g
A2244100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A224425G Supplier TCI America Supplier No. A224425G

Chemical Identifiers

CAS 6358-08-3
Molecular Formula C6H5ClN2O3
Molecular Weight (g/mol) 188.57
MDL Number MFCD00035925
InChI Key MHAFRUMLQZZSIN-UHFFFAOYSA-N
PubChem CID 4348294
IUPAC Name 2-amino-4-chloro-6-nitrophenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)N)Cl

Specifications

Melting Point 152°C
Quantity 25 g
Synonym unii-13d00yq1u0, 6-amino-4-chloro-2-nitrophenol, 2-amino-4-chloro-6-nitro-phenol, 2-amino-4-chloro-6-nitro phenol, phenol, 2-amino-4-chloro-6-nitro, acmc-2097jj, 4-phenylethyl isothiocyanate, 4-chlor-6-nitro-2-aminophenol, 5-chloro-2-hydroxy-3-nitroaniline, phenol,2-amino-4-chloro-6-nitro
Formula Weight 188.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-nitrophenol
EINECSNumber (3)-0939
TSCA No