missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methoxypyrimidine, 95%, Thermo Scientific™

Catalog No. p-54937
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347090250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC347090250 Supplier Thermo Scientific Chemicals Supplier No. 347090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5734-64-5
Molecular Formula C5H6ClN3O
Molecular Weight (g/mol) 159.57
MDL Number MFCD00233535
InChI Key VFEYBTFCBZMBAU-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
PubChem CID 79811
IUPAC Name 4-chloro-6-methoxypyrimidin-2-amine
SMILES COC1=CC(Cl)=NC(N)=N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 169°C to 176°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Formula Weight 159.57
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber UV6260335
TSCA TSCA

RUO – Research Use Only