missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-6-hydroxypyrimidine 98.0+%, TCI America™
$158.44 - $531.25
Chemical Identifiers
| CAS | 1194-21-4 |
|---|---|
| Molecular Formula | C4H4ClN3O |
| Molecular Weight (g/mol) | 145.546 |
| MDL Number | MFCD00006093 |
| InChI Key | VBWACOJLJYUFKJ-UHFFFAOYSA-N |
| Synonym | 2-Amino-4-chloro-6-pyrimidinol |
| PubChem CID | 70948 |
| IUPAC Name | 2-amino-6-chloro-1H-pyrimidin-4-one |
| SMILES | C1=C(NC(=NC1=O)N)Cl |
Chemical Identifiers
| 1194-21-4 | |
| 145.546 | |
| VBWACOJLJYUFKJ-UHFFFAOYSA-N | |
| 70948 | |
| C1=C(NC(=NC1=O)N)Cl |
| C4H4ClN3O | |
| MFCD00006093 | |
| 2-Amino-4-chloro-6-pyrimidinol | |
| 2-amino-6-chloro-1H-pyrimidin-4-one |
Specifications
| 1194-21-4 | |
| MFCD00006093 | |
| 2-Amino-4-chloro-6-pyrimidinol | |
| C1=C(NC(=NC1=O)N)Cl | |
| 145.546 | |
| 145.55 | |
| Crystalline Powder |
| C4H4ClN3O | |
| 5 g | |
| VBWACOJLJYUFKJ-UHFFFAOYSA-N | |
| 2-amino-6-chloro-1H-pyrimidin-4-one | |
| 70948 | |
| ≥98.0% (HPLC,T) | |
| 2-Amino-4-chloro-6-hydroxypyrimidine |
Safety and Handling
RTECSNumber : UW5995000
TSCA : No