missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-hydroxypyrimidine 98.0+%, TCI America™

Supplier:  TCI America A11095G

 View more versions of this product

Catalog No. A11095G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Amino-4-chloro-6-hydroxypyrimidine
C4H4ClN3O
5 g
VBWACOJLJYUFKJ-UHFFFAOYSA-N
2-amino-6-chloro-1H-pyrimidin-4-one
70948
≥98.0% (HPLC,T)
1194-21-4
MFCD00006093
2-Amino-4-chloro-6-pyrimidinol
C1=C(NC(=NC1=O)N)Cl
145.546
145.55
Crystalline Powder

Chemical Identifiers

1194-21-4
145.546
VBWACOJLJYUFKJ-UHFFFAOYSA-N
70948
C1=C(NC(=NC1=O)N)Cl
C4H4ClN3O
MFCD00006093
2-Amino-4-chloro-6-pyrimidinol
2-amino-6-chloro-1H-pyrimidin-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UW5995000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions