missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-5-nitrophenol 98.0+%, TCI America™
$73.16 - $506.33
Chemical Identifiers
| CAS | 6358-07-2 |
|---|---|
| Molecular Formula | C6H5ClN2O3 |
| Molecular Weight (g/mol) | 188.57 |
| MDL Number | MFCD00010300 |
| InChI Key | ZARYBZGMUVAJMK-UHFFFAOYSA-N |
| Synonym | 5-Chloro-2-hydroxy-4-nitroaniline |
| PubChem CID | 3894848 |
| IUPAC Name | 2-amino-4-chloro-5-nitrophenol |
| SMILES | C1=C(C(=CC(=C1Cl)[N+](=O)[O-])O)N |
Chemical Identifiers
| 6358-07-2 | |
| 188.57 | |
| ZARYBZGMUVAJMK-UHFFFAOYSA-N | |
| 3894848 | |
| C1=C(C(=CC(=C1Cl)[N+](=O)[O-])O)N |
| C6H5ClN2O3 | |
| MFCD00010300 | |
| 5-Chloro-2-hydroxy-4-nitroaniline | |
| 2-amino-4-chloro-5-nitrophenol |
Specifications
| 6358-07-2 | |
| C6H5ClN2O3 | |
| 25 g | |
| ZARYBZGMUVAJMK-UHFFFAOYSA-N | |
| 2-amino-4-chloro-5-nitrophenol | |
| 3894848 | |
| ≥98.0% (T) | |
| 2-Amino-4-chloro-5-nitrophenol |
| 220°C | |
| MFCD00010300 | |
| 5-Chloro-2-hydroxy-4-nitroaniline | |
| C1=C(C(=CC(=C1Cl)[N+](=O)[O-])O)N | |
| 188.57 | |
| 188.57 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SJ5736000
TSCA : Yes