missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile 98.0+%, TCI America™
$128.62 - $391.44
Chemical Identifiers
| CAS | 104366-23-6 |
|---|---|
| Molecular Formula | C6H3ClN2OS |
| Molecular Weight (g/mol) | 186.613 |
| MDL Number | MFCD03788265 |
| InChI Key | XIZSUYSWKGAOCN-UHFFFAOYSA-N |
| Synonym | 2-Amino-4-chloro-3-cyano-5-formylthiophene |
| PubChem CID | 2737612 |
| IUPAC Name | 2-amino-4-chloro-5-formylthiophene-3-carbonitrile |
| SMILES | C(=O)C1=C(C(=C(S1)N)C#N)Cl |
Chemical Identifiers
| 104366-23-6 | |
| 186.613 | |
| XIZSUYSWKGAOCN-UHFFFAOYSA-N | |
| 2737612 | |
| C(=O)C1=C(C(=C(S1)N)C#N)Cl |
| C6H3ClN2OS | |
| MFCD03788265 | |
| 2-Amino-4-chloro-3-cyano-5-formylthiophene | |
| 2-amino-4-chloro-5-formylthiophene-3-carbonitrile |
Specifications
| 104366-23-6 | |
| C6H3ClN2OS | |
| 1 g | |
| 2-Amino-4-chloro-3-cyano-5-formylthiophene | |
| C(=O)C1=C(C(=C(S1)N)C#N)Cl | |
| 186.613 | |
| 186.61 | |
| Crystalline Powder |
| 269°C | |
| MFCD03788265 | |
| 2923 | |
| XIZSUYSWKGAOCN-UHFFFAOYSA-N | |
| 2-amino-4-chloro-5-formylthiophene-3-carbonitrile | |
| 2737612 | |
| ≥98.0% (GC) | |
| 2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator