accessibility menu, dialog, popup

UserName

OSH2NNCl

CAS RN 104366-23-6

IUPAC Name: 2-amino-4-chloro-5-formylthiophene-3-carbonitrile
Synonyms: 2-amino-4-chloro-5-formylthiophene-3-carbonitrile
Molecular Weight (g/mol): 186.61
Molecular Formula: C6H3ClN2OS
InChi Key: XIZSUYSWKGAOCN-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C(Cl)=C(S1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results