missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 269°C |
| Quantity | 5 g |
| UN Number | 2923 |
| Formula Weight | 186.61 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |