missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile 98.0+%, TCI America™

Catalog No. A28631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28631G 1 g
A28635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28631G Supplier TCI America Supplier No. A28631G
Only null left

Chemical Identifiers

CAS 104366-23-6
Molecular Formula C6H3ClN2OS
Molecular Weight (g/mol) 186.613
MDL Number MFCD03788265
InChI Key XIZSUYSWKGAOCN-UHFFFAOYSA-N
Synonym 2-Amino-4-chloro-3-cyano-5-formylthiophene
PubChem CID 2737612
IUPAC Name 2-amino-4-chloro-5-formylthiophene-3-carbonitrile
SMILES C(=O)C1=C(C(=C(S1)N)C#N)Cl

Specifications

Melting Point 269°C
Quantity 1 g
UN Number 2923
Formula Weight 186.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile
TSCA No
Recommended Storage Refrigerator