missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromophenol 98.0+%, TCI America™

Catalog No. A22715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22715G 5 g
A227125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22715G Supplier TCI America Supplier No. A22715G

Chemical Identifiers

CAS 40925-68-6
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00235171
InChI Key JHRIPENGTGSNPJ-UHFFFAOYSA-N
Synonym phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p
PubChem CID 351840
IUPAC Name 2-amino-4-bromophenol
SMILES NC1=CC(Br)=CC=C1O

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-bromophenol