missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-bromoacetophenone Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118198
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30301G 1 g
A30305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30301G Supplier TCI America Supplier No. A30301G
Only null left

Chemical Identifiers

CAS 5467-72-1
Molecular Formula C8H9BrClNO
Molecular Weight (g/mol) 250.52
MDL Number MFCD00051998
InChI Key ROAVTVXTYFSQEA-UHFFFAOYSA-N
Synonym 4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
PubChem CID 2798216
IUPAC Name 2-amino-1-(4-bromophenyl)ethanone;hydrochloride
SMILES C1=CC(=CC=C1C(=O)CN)Br.Cl

Specifications

Melting Point 285°C
Color White-Yellow
Quantity 1 g
Formula Weight 250.52
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4′-bromoacetophenone Hydrochloride
RTECSNumber AM5840000
TSCA No