accessibility menu, dialog, popup

UserName

ClNH3OBr

CAS RN 5467-72-1

IUPAC Name: 2-(4-bromophenyl)-2-oxoethan-1-aminium chloride
Synonyms: 2-(4-bromophenyl)-2-oxoethanaminium chloride
Molecular Weight (g/mol): 250.52
Molecular Formula: C8H9BrClNO
InChi Key: ROAVTVXTYFSQEA-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)