missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-bromoacetophenone hydrochloride, 98%

Catalog No. p-7044055
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0429204 2 g
AAL0429209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0429204 Supplier Thermo Scientific Chemicals Supplier No. L0429204
Only null left

CAS: 5467-72-1 | C8H9BrClNO | 250.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5467-72-1
Molecular Formula C8H9BrClNO
Molecular Weight (g/mol) 250.52
MDL Number MFCD00051998
InChI Key ROAVTVXTYFSQEA-UHFFFAOYSA-N
Synonym 4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
PubChem CID 2798216
IUPAC Name 2-amino-1-(4-bromophenyl)ethanone;hydrochloride
SMILES C1=CC(=CC=C1C(=O)CN)Br.Cl

Specifications

Melting Point 280°C to 283°C
Quantity 2 g
Beilstein 3630035
Formula Weight 250.53
Percent Purity 98%
Chemical Name or Material 2-Amino-4'-bromoacetophenone hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-778-3
RTECSNumber AM5840000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only