missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-bromoacetophenone hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5467-72-1
Molecular Formula C8H8BrNO·ClH
Molecular Weight (g/mol) 250.52
MDL Number MFCD00051998
InChI Key ROAVTVXTYFSQEA-UHFFFAOYSA-N
Synonym 4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
PubChem CID 2798216
IUPAC Name 2-amino-1-(4-bromophenyl)ethanone;hydrochloride
SMILES C1=CC(=CC=C1C(=O)CN)Br.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC427060050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5467-72-1
250.52
ROAVTVXTYFSQEA-UHFFFAOYSA-N
2798216
C1=CC(=CC=C1C(=O)CN)Br.Cl
C8H8BrNO·ClH
MFCD00051998
4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
2-amino-1-(4-bromophenyl)ethanone;hydrochloride

Specifications

5467-72-1
100.0
C8H8BrNO·ClH
MFCD00051998
Solubility in water: soluble
C1=CC(=CC=C1C(=O)CN)Br.Cl
250.52
250.52
2-Amino-4′-bromoacetophenone hydrochloride
96.0
96% min. (HPLC)
BrC6H4C(O)CH2NH2·HCl
4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
ROAVTVXTYFSQEA-UHFFFAOYSA-N
2-amino-1-(4-bromophenyl)ethanone;hydrochloride
2798216
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-778-3

SDS
Documents

Documents

RUO – Research Use Only