missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethylthiazole Hydrochloride 98.0+%, TCI America™

$89.48 - $249.78

Chemical Identifiers

CAS 71574-33-9
Molecular Formula C5H9ClN2S
Molecular Weight (g/mol) 164.651
MDL Number MFCD00012711
InChI Key XYTUTNQRQLAZLK-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
PubChem CID 16211387
IUPAC Name 4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride
SMILES CC1=C(SC(=N1)N)C.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A02921G
View Documents
TCI America
A02921G
1 g
Each for $89.48
Only null left
 
A02925G
View Documents
TCI America
A02925G
5 g
Each for $249.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

71574-33-9
164.651
XYTUTNQRQLAZLK-UHFFFAOYSA-N
16211387
CC1=C(SC(=N1)N)C.Cl
C5H9ClN2S
MFCD00012711
4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride

Specifications

71574-33-9
C5H9ClN2S
1 g
XYTUTNQRQLAZLK-UHFFFAOYSA-N
4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride
16211387
≥98.0% (T)
2-Amino-4,5-dimethylthiazole Hydrochloride
White
MFCD00012711
4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
CC1=C(SC(=N1)N)C.Cl
164.651
164.65
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents