missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethylthiazole Hydrochloride 98.0+%, TCI America™

Catalog No. A02925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02925G 5 g
A02921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A02925G Supplier TCI America Supplier No. A02925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71574-33-9
Molecular Formula C5H9ClN2S
Molecular Weight (g/mol) 164.651
MDL Number MFCD00012711
InChI Key XYTUTNQRQLAZLK-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
PubChem CID 16211387
IUPAC Name 4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride
SMILES CC1=C(SC(=N1)N)C.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 164.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,5-dimethylthiazole Hydrochloride