missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethylthiazole hydrochloride, 97%

Catalog No. AAH5987809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5987809 10 g
AAH5987818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5987809 Supplier Thermo Scientific Chemicals Supplier No. H5987809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71574-33-9 | C5H9ClN2S | 164.651 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71574-33-9
Molecular Formula C5H9ClN2S
Molecular Weight (g/mol) 164.651
MDL Number MFCD00012711
InChI Key XYTUTNQRQLAZLK-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazol-2-amine hydrochloride, 2-amino-4,5-dimethylthiazole hydrochloride, 4,5-dimethyl-1,3-thiazol-2-amine hydrochloride, dimethyl-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 4,5-dimethyl-, monohydrochloride, acmc-209oj8, ksc496i8r, 4,5-dimethylthiazol-2-amine hcl salt, 4,5-dimethylthiazol-2-amine hcl, 2-amino-4,5-dimethyl thiazole hydrochloride
PubChem CID 16211387
IUPAC Name 4,5-dimethyl-1,3-thiazol-2-amine;hydrochloride
SMILES CC1=C(SC(=N1)N)C.Cl

Specifications

Melting Point 273°C to 274°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 164.66
Percent Purity 98%
Chemical Name or Material 2-Amino-4,5-dimethylthiazole hydrochloride
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only