missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7044398
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0620514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0620514 Supplier Thermo Scientific Chemicals Supplier No. L0620514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5653-40-7 | Molecular Formula: C9H11NO4 | Molecular Weight: 197.19 g/mol

2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Methanol and Water

Notes
Incompatible with bases and oxidizing agents. Store at -20°C.

Chemical Identifiers

CAS 5653-40-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00011671
InChI Key HJVAVGOPTDJYOJ-UHFFFAOYSA-N
Synonym 4,5-dimethoxyanthranilic acid, 6-aminoveratric acid, benzoic acid, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy benzoic acid, 2-amino-4,5-dimethoxy-benzoic acid, 6-aminoveratricacid, zlchem 238, pubchem3110, acmc-1b03r, ksc269i1n
PubChem CID 79736
IUPAC Name 2-amino-4,5-dimethoxybenzoic acid
SMILES COC1=CC(N)=C(C=C1OC)C(O)=O

Specifications

Melting Point ∽155°C to 158°C (decomposition)
Quantity 25 g
Beilstein 782814
Solubility Information Soluble in Methanol and Water
Formula Weight 197.19
Percent Purity 98%
Chemical Name or Material 2-Amino-4,5-dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-095-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only