missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,4'-dichlorodiphenyl Ether 98.0+%, TCI America™
$190.13 - $619.05
Chemical Identifiers
| CAS | 121-27-7 |
|---|---|
| Molecular Formula | C12H9Cl2NO |
| Molecular Weight (g/mol) | 254.11 |
| MDL Number | MFCD00027391 |
| InChI Key | WLJSUJOESWTGEX-UHFFFAOYSA-N |
| Synonym | 5-Chloro-2-(4-chlorophenoxy)aniline |
| PubChem CID | 67135 |
| IUPAC Name | 5-chloro-2-(4-chlorophenoxy)aniline |
| SMILES | C1=CC(=CC=C1OC2=C(C=C(C=C2)Cl)N)Cl |
Chemical Identifiers
| 121-27-7 | |
| 254.11 | |
| WLJSUJOESWTGEX-UHFFFAOYSA-N | |
| 67135 | |
| C1=CC(=CC=C1OC2=C(C=C(C=C2)Cl)N)Cl |
| C12H9Cl2NO | |
| MFCD00027391 | |
| 5-Chloro-2-(4-chlorophenoxy)aniline | |
| 5-chloro-2-(4-chlorophenoxy)aniline |
Specifications
| 121-27-7 | |
| 234°C | |
| MFCD00027391 | |
| 5-Chloro-2-(4-chlorophenoxy)aniline | |
| C1=CC(=CC=C1OC2=C(C=C(C=C2)Cl)N)Cl | |
| 254.11 | |
| 254.11 | |
| Crystalline Powder |
| 64°C | |
| C12H9Cl2NO | |
| 5 g | |
| WLJSUJOESWTGEX-UHFFFAOYSA-N | |
| 5-chloro-2-(4-chlorophenoxy)aniline | |
| 67135 | |
| ≥98.0% (GC,T) | |
| 2-Amino-4,4′-dichlorodiphenyl Ether |
Safety and Handling
TSCA : Yes