missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,4'-dichlorodiphenyl Ether 98.0+%, TCI America™

Catalog No. A22255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22255G 5 g
A222525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22255G Supplier TCI America Supplier No. A22255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-27-7
Molecular Formula C12H9Cl2NO
Molecular Weight (g/mol) 254.11
MDL Number MFCD00027391
InChI Key WLJSUJOESWTGEX-UHFFFAOYSA-N
Synonym 5-Chloro-2-(4-chlorophenoxy)aniline
PubChem CID 67135
IUPAC Name 5-chloro-2-(4-chlorophenoxy)aniline
SMILES C1=CC(=CC=C1OC2=C(C=C(C=C2)Cl)N)Cl

Specifications

Melting Point 64°C
Boiling Point 234°C
Quantity 5 g
Formula Weight 254.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,4′-dichlorodiphenyl Ether