missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-cyanophenyl)thiazole, 97%

Catalog No. p-7036551
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54861MD 250 mg
AAH5486103 1 g
AAH5486106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54861MD Supplier Thermo Scientific Chemicals Supplier No. H54861MD
Only null left

CAS: 436151-85-8 | C10H7N3S | 201.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 436151-85-8
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.25
MDL Number MFCD03407828
InChI Key KKMZKOIZTSRIEM-UHFFFAOYSA-N
Synonym 4-2-amino-1,3-thiazol-4-yl benzonitrile, 4-2-aminothiazol-4-yl benzonitrile, 4-2-amino-thiazol-4-yl-benzonitrile, benzonitrile, 4-2-amino-4-thiazolyl, acmc-1aqww, 2-amino-4-4-cyanophenyl thiazole, 4-4-cyanophenyl-2-aminothiazole, 4-2-amino-4-thiazolyl benzonitrile, 4-2-aminothiazole-4-yl benzonitrile, 4-2-amino-thiazol-4-yl benzonitrile
PubChem CID 2049589
IUPAC Name 4-(2-amino-1,3-thiazol-4-yl)benzonitrile
SMILES NC1=NC(=CS1)C1=CC=C(C=C1)C#N

Specifications

Quantity 250 mg
Beilstein 15029305
Formula Weight 201.25
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(4-cyanophenyl)thiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only