CAS RN 436151-85-8
IUPAC Name:
4-(2-amino-1,3-thiazol-4-yl)benzonitrile
Synonyms:
4-(2-amino-1,3-thiazol-4-yl)benzonitrile
Molecular Weight (g/mol):
201.25
Molecular Formula:
C10H7N3S
InChi Key:
KKMZKOIZTSRIEM-UHFFFAOYSA-N
SMILES:
NC1=NC(=CS1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Promotions Available
2-Amino-4-(4-cyanophenyl)thiazole, 97%
2-Amino-4-(4-cyanophenyl)thiazole, 97%
CAS: 436151-85-8 Molecular Formula: C10H7N3S Molecular Weight (g/mol): 201.25 MDL Number: MFCD03407828 InChI Key: KKMZKOIZTSRIEM-UHFFFAOYSA-N Synonym: 4-2-amino-1,3-thiazol-4-yl benzonitrile,4-2-aminothiazol-4-yl benzonitrile,4-2-amino-thiazol-4-yl-benzonitrile,benzonitrile, 4-2-amino-4-thiazolyl,acmc-1aqww,2-amino-4-4-cyanophenyl thiazole,4-4-cyanophenyl-2-aminothiazole,4-2-amino-4-thiazolyl benzonitrile,4-2-aminothiazole-4-yl benzonitrile,4-2-amino-thiazol-4-yl benzonitrile PubChem CID: 2049589 IUPAC Name: 4-(2-amino-1,3-thiazol-4-yl)benzonitrile SMILES: NC1=NC(=CS1)C1=CC=C(C=C1)C#N