missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,4,4-trifluorobutyric acid, 97%

CAS: 15959-93-0 | C4H6F3NO2 | 157.092 g/mol

Supplier:  Thermo Scientific Chemicals L13131ME

 View more versions of this product

Catalog No. AAL13131ME


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-4,4,4-trifluorobutyric acid
∽250°C (decomposition)
MFCD00041417
1769355
AQPCXCOPDSEKQT-UHFFFAOYSA-N
2-amino-4,4,4-trifluorobutanoic acid
353083
97%
15959-93-0
C4H6F3NO2
500 mg
2-amino-4,4,4-trifluorobutyric acid, butanoic acid, 2-amino-4,4,4-trifluoro, s-2-amino-4,4,4-trifluorobutyric acid, 3-trifluoromethylalanine, 2-amino-4,4,4-trifluoro-butyric acid, butanoicacid,2-amino-4,4,4-trifluoro, butanoic acid,2-amino-4,4,4-trifluoro, 2-amino-4,4,4-trifluoro-n-butyric acid hydrochlori
C(C(C(=O)O)N)C(F)(F)F
157.092
157.09

Chemical Identifiers

15959-93-0
157.092
AQPCXCOPDSEKQT-UHFFFAOYSA-N
2-amino-4,4,4-trifluorobutanoic acid
C4H6F3NO2
MFCD00041417
353083
C(C(C(=O)O)N)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only