missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,4,4-trifluorobutyric acid, 97%

Catalog No. p-7046102
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13131MC 100 mg
AAL13131ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13131MC Supplier Thermo Scientific Chemicals Supplier No. L13131MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15959-93-0 | C4H6F3NO2 | 157.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15959-93-0
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.092
MDL Number MFCD00041417
InChI Key AQPCXCOPDSEKQT-UHFFFAOYSA-N
PubChem CID 353083
IUPAC Name 2-amino-4,4,4-trifluorobutanoic acid
SMILES C(C(C(=O)O)N)C(F)(F)F

Specifications

Melting Point ∽250°C (decomposition)
Quantity 100 mg
Beilstein 1769355
Synonym 2-amino-4,4,4-trifluorobutyric acid, butanoic acid, 2-amino-4,4,4-trifluoro, s-2-amino-4,4,4-trifluorobutyric acid, 3-trifluoromethylalanine, 2-amino-4,4,4-trifluoro-butyric acid, butanoicacid,2-amino-4,4,4-trifluoro, butanoic acid,2-amino-4,4,4-trifluoro, 2-amino-4,4,4-trifluoro-n-butyric acid hydrochlori
Formula Weight 157.09
Percent Purity 97%
Chemical Name or Material 2-Amino-4,4,4-trifluorobutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only