missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(3,4-dichlorophenyl)thiazole 98.0+%, TCI America™

Catalog No. A29061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29061G 1 g
A29065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29061G Supplier TCI America Supplier No. A29061G
Only null left

Chemical Identifiers

CAS 39893-80-6
Molecular Formula C9H6Cl2N2S
Molecular Weight (g/mol) 245.12
MDL Number MFCD00466323
InChI Key UWAXMUSNRIUHRQ-UHFFFAOYSA-N
Synonym 4-(3,4-Dichlorophenyl)-2-thiazolamine
PubChem CID 618536
IUPAC Name 4-(3,4-dichlorophenyl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 163°C
Color Yellow
Quantity 1 g
Formula Weight 245.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(3,4-dichlorophenyl)thiazole