missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-chloro-3-pyridyl)pyrimidine, 97%, Thermo Scientific™

$600.21 - $600.21

Chemical Identifiers

CAS 870221-49-1
Molecular Formula C9H7ClN4
Molecular Weight (g/mol) 206.633
MDL Number MFCD18157687
InChI Key DKCVVHMYFZBDAK-UHFFFAOYSA-N
PubChem CID 53396335
IUPAC Name 4-(2-chloropyridin-3-yl)pyrimidin-2-amine
SMILES C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5495703
View Documents
Thermo Scientific Chemicals
H5495703
1 g
Each for $600.21
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as primary and secondary intermediate.

Notes
Incompatible with strong oxidizing agents, bases, acids. Keep in room temperature.
Specifications

Chemical Identifiers

870221-49-1
206.633
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
C9H7ClN4
MFCD18157687
53396335
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N

Specifications

870221-49-1
MFCD18157687
14484335
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
53396335
97%
C9H7ClN4
1 g
4-2-chloropyridin-3-yl pyrimidin-2-amine, chloropyridinylpyrimidinamine, 2-pyrimidinamine,4-2-chloro-3-pyridinyl, 2-pyrimidinamine, 4-2-chloro-3-pyridinyl
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
206.633
206.63
2-Amino-4-(2-chloro-3-pyridyl)pyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'