accessibility menu, dialog, popup

UserName

CAS RN 870221-49-1

IUPAC Name: 4-(2-chloropyridin-3-yl)pyrimidin-2-amine
Synonyms: 4-(2-chloropyridin-3-yl)pyrimidin-2-amine
Molecular Weight (g/mol): 206.63
Molecular Formula: C9H7ClN4
InChi Key: DKCVVHMYFZBDAK-UHFFFAOYSA-N
SMILES: NC1=NC=CC(=N1)C1=C(Cl)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results