missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2,4-difluorophenyl)thiazole 98.0+%, TCI America™

Catalog No. A30295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30295G 5 g
A30291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30295G Supplier TCI America Supplier No. A30295G

Chemical Identifiers

CAS 105512-80-9
Molecular Formula C9H6F2N2S
Molecular Weight (g/mol) 212.22
MDL Number MFCD00472891
InChI Key HKUPOLYVZQLXKP-UHFFFAOYSA-N
Synonym 4-2,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-2,4-difluorophenyl thiazole, 4-2,4-difluorophenyl thiazol-2-amine, 2-amino-4-2,4-difluorophenyl-1,3-thiazole, 4-2,4-difluorophenyl thiazol-2-ylamine, 4-2,4-difluoro-phenyl-thiazol-2-ylamine, 4-2,4-difluorophenyl-1,3-thiazole-2-ylamine, acmc-1bro0, cbmicro_034355, 2-thiazolamine,4-2,4-difluorophenyl
PubChem CID 611370
IUPAC Name 4-(2,4-difluorophenyl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=C(F)C=C(F)C=C1

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(2,4-difluorophenyl)thiazole