CAS RN 105512-80-9
IUPAC Name:
4-(2,4-difluorophenyl)-1,3-thiazol-2-amine
Synonyms:
4-(2,4-difluorophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol):
212.22
Molecular Formula:
C9H6F2N2S
InChi Key:
HKUPOLYVZQLXKP-UHFFFAOYSA-N
SMILES:
NC1=NC(=CS1)C1=C(F)C=C(F)C=C1
1
–
1
of
1
results
1
2-Amino-4-(2,4-difluorophenyl)thiazole 98.0+%, TCI America™
CAS: 105512-80-9 Molecular Formula: C9H6F2N2S Molecular Weight (g/mol): 212.22 MDL Number: MFCD00472891 InChI Key: HKUPOLYVZQLXKP-UHFFFAOYSA-N Synonym: 4-2,4-difluorophenyl-1,3-thiazol-2-amine,2-amino-4-2,4-difluorophenyl thiazole,4-2,4-difluorophenyl thiazol-2-amine,2-amino-4-2,4-difluorophenyl-1,3-thiazole,4-2,4-difluorophenyl thiazol-2-ylamine,4-2,4-difluoro-phenyl-thiazol-2-ylamine,4-2,4-difluorophenyl-1,3-thiazole-2-ylamine,acmc-1bro0,cbmicro_034355,2-thiazolamine,4-2,4-difluorophenyl PubChem CID: 611370 IUPAC Name: 4-(2,4-difluorophenyl)-1,3-thiazol-2-amine SMILES: NC1=NC(=CS1)C1=C(F)C=C(F)C=C1