missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(1-naphthyl)thiazole 97.0+%, TCI America™

Catalog No. A28885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28885G 5 g
A28881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28885G Supplier TCI America Supplier No. A28885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56503-96-9
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.30
MDL Number MFCD00236016
InChI Key NBQUWOCIFFHZTM-UHFFFAOYSA-N
Synonym 2-amino-4-1-naphthyl thiazole, 4-1-naphthalenyl-2-thiazolamine, 4-naphthalen-1-yl-thiazol-2-ylamine, 2-thiazolamine, 4-1-naphthalenyl, 4-naphthalen-1-yl-1,3-thiazol-2-amine, 4-naphthalen-1-yl thiazol-2-amine, 4-naphthalen-1-yl-thiazol-2-yl-amine, 4-1-naphthyl-1,3-thiazol-2-amine, 2-amino-4-alpha-naphthylthiazole, 2-amino-4-1-naphthalenyl-1,3-thiazole
PubChem CID 151342
IUPAC Name 4-(naphthalen-1-yl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 158°C
Quantity 5 g
Formula Weight 226.30
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(1-naphthyl)thiazole
RTECSNumber XJ1990000
TSCA No