accessibility menu, dialog, popup

UserName

NH2SN

CAS RN 56503-96-9

IUPAC Name: 4-(naphthalen-1-yl)-1,3-thiazol-2-amine
Synonyms: 4-(naphthalen-1-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 226.3
Molecular Formula: C13H10N2S
InChi Key: NBQUWOCIFFHZTM-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)