missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(1-naphthyl)thiazole, 97%

Catalog No. p-7027261
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2388203 1 g
AAB2388206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2388203 Supplier Thermo Scientific Chemicals Supplier No. B2388203
Only null left

CAS: 56503-96-9 | C13H10N2S | 226.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56503-96-9
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.30
MDL Number MFCD00236016
InChI Key NBQUWOCIFFHZTM-UHFFFAOYSA-N
Synonym 2-amino-4-1-naphthyl thiazole, 4-1-naphthalenyl-2-thiazolamine, 4-naphthalen-1-yl-thiazol-2-ylamine, 2-thiazolamine, 4-1-naphthalenyl, 4-naphthalen-1-yl-1,3-thiazol-2-amine, 4-naphthalen-1-yl thiazol-2-amine, 4-naphthalen-1-yl-thiazol-2-yl-amine, 4-1-naphthyl-1,3-thiazol-2-amine, 2-amino-4-alpha-naphthylthiazole, 2-amino-4-1-naphthalenyl-1,3-thiazole
PubChem CID 151342
IUPAC Name 4-naphthalen-1-yl-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 155°C to 158°C
Quantity 1 g
Beilstein 164858
Formula Weight 226.3
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(1-naphthyl)thiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber XJ1990000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only