missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-pyridinemethanol 96.0+%, TCI America™
$91.33 - $311.30
Chemical Identifiers
| CAS | 23612-57-9 |
|---|---|
| Molecular Formula | C6H8N2O |
| Molecular Weight (g/mol) | 124.14 |
| MDL Number | MFCD05663510 |
| InChI Key | FEIACFYXEWBKHU-UHFFFAOYSA-N |
| Synonym | 2-aminopyridin-3-yl methanol, 2-aminopyridine-3-methanol, 2-amino-3-hydroxymethylpyridine, 2-amino-pyridin-3-yl-methanol, 2-amino-3-pyridinyl methanol, 2-amino-3-pyridinemethanol, 2-amino-3-hydroxymethyl pyridine, 3-pyridinemethanol, 2-amino, 2-aminopyridine-3-yl-methanol |
| PubChem CID | 11159325 |
| IUPAC Name | (2-aminopyridin-3-yl)methanol |
| SMILES | NC1=NC=CC=C1CO |
Chemical Identifiers
| 23612-57-9 | |
| 124.14 | |
| FEIACFYXEWBKHU-UHFFFAOYSA-N | |
| 11159325 | |
| NC1=NC=CC=C1CO |
| C6H8N2O | |
| MFCD05663510 | |
| 2-aminopyridin-3-yl methanol, 2-aminopyridine-3-methanol, 2-amino-3-hydroxymethylpyridine, 2-amino-pyridin-3-yl-methanol, 2-amino-3-pyridinyl methanol, 2-amino-3-pyridinemethanol, 2-amino-3-hydroxymethyl pyridine, 3-pyridinemethanol, 2-amino, 2-aminopyridine-3-yl-methanol | |
| (2-aminopyridin-3-yl)methanol |
Specifications
| 23612-57-9 | |
| Yellow | |
| C6H8N2O | |
| 1 g | |
| FEIACFYXEWBKHU-UHFFFAOYSA-N | |
| (2-aminopyridin-3-yl)methanol | |
| 11159325 | |
| ≥96.0% (GC) | |
| 2-Amino-3-pyridinemethanol |
| 67°C | |
| 279°C | |
| MFCD05663510 | |
| 2-aminopyridin-3-yl methanol, 2-aminopyridine-3-methanol, 2-amino-3-hydroxymethylpyridine, 2-amino-pyridin-3-yl-methanol, 2-amino-3-pyridinyl methanol, 2-amino-3-pyridinemethanol, 2-amino-3-hydroxymethyl pyridine, 3-pyridinemethanol, 2-amino, 2-aminopyridine-3-yl-methanol | |
| NC1=NC=CC=C1CO | |
| 124.14 | |
| 124.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No