CAS RN 23612-57-9
CAS RN 23612-57-9
IUPAC Name:
(2-aminopyridin-3-yl)methanol
Synonyms:
(2-aminopyridin-3-yl)methanol
Molecular Weight (g/mol):
124.14
Molecular Formula:
C6H8N2O
InChi Key:
FEIACFYXEWBKHU-UHFFFAOYSA-N
SMILES:
NC1=NC=CC=C1CO
1
–
1
of
1
results
1
2-Amino-3-pyridinemethanol 96.0+%, TCI America™
CAS: 23612-57-9 Molecular Formula: C6H8N2O Molecular Weight (g/mol): 124.14 MDL Number: MFCD05663510 InChI Key: FEIACFYXEWBKHU-UHFFFAOYSA-N Synonym: 2-aminopyridin-3-yl methanol,2-aminopyridine-3-methanol,2-amino-3-hydroxymethylpyridine,2-amino-pyridin-3-yl-methanol,2-amino-3-pyridinyl methanol,2-amino-3-pyridinemethanol,2-amino-3-hydroxymethyl pyridine,3-pyridinemethanol, 2-amino,2-aminopyridine-3-yl-methanol PubChem CID: 11159325 IUPAC Name: (2-aminopyridin-3-yl)methanol SMILES: NC1=NC=CC=C1CO