missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitrobenzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 606-18-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.13
InChI Key JJPIVRWTAGQTPQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
PubChem CID 219633
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC443180100
SDS View Documents
Thermo Scientific Chemicals
443180100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

606-18-8
182.13
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
C7H6N2O4
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
219633

Specifications

606-18-8
C7H6N2O4
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
219633
97%
96% min. (HPLC)
10g
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
182.13
182.13
2-Amino-3-nitrobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only