missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3'-nitroacetophenone hydrochloride, 98%

Catalog No. p-7081984
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1220503 1 g
AAA1220506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1220503 Supplier Thermo Scientific Chemicals Supplier No. A1220503
Only null left

CAS: 36765-84-1 | C8H9N2O3 | 181.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36765-84-1
Molecular Formula C8H9N2O3
Molecular Weight (g/mol) 181.17
MDL Number MFCD00051997
InChI Key NXCCDPOCDBAGFX-UHFFFAOYSA-O
Synonym 3-nitrophenacylamine hydrochloride, 2-amino-1-3-nitrophenyl ethanone hydrochloride, 2-amino-3'-nitroacetophenone hydrochloride, 2-amino-1-3-nitrophenyl ethan-1-one hydrochloride, acmc-20amzi, 2-amine-3'-nitroacetophenone hcl, 2-amino-1-3-nitrophenyl ethanone hcl salt, 2-amino-3-nitroacetophenone hydrochloride
PubChem CID 2774374
IUPAC Name 2-amino-1-(3-nitrophenyl)ethanone;hydrochloride
SMILES [NH3+]CC(=O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 210°C to 212°C (decomposition)
Quantity 1 g
UN Number UN1759
Formula Weight 216.62
Percent Purity 98%
Chemical Name or Material 2-Amino-3'-nitroacetophenone hydrochloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only