CAS RN 36765-84-1
IUPAC Name:
2-(3-nitrophenyl)-2-oxoethan-1-aminium
Synonyms:
2-(3-nitrophenyl)-2-oxoethanaminium
Molecular Weight (g/mol):
181.17
Molecular Formula:
C8H9N2O3
InChi Key:
NXCCDPOCDBAGFX-UHFFFAOYSA-O
SMILES:
[NH3+]CC(=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2-Amino-3'-nitroacetophenone hydrochloride, 98%
2-Amino-3'-nitroacetophenone hydrochloride, 98%
CAS: 36765-84-1 Molecular Formula: C8H9N2O3 Molecular Weight (g/mol): 181.17 MDL Number: MFCD00051997 InChI Key: NXCCDPOCDBAGFX-UHFFFAOYSA-O Synonym: 3-nitrophenacylamine hydrochloride,2-amino-1-3-nitrophenyl ethanone hydrochloride,2-amino-3'-nitroacetophenone hydrochloride,2-amino-1-3-nitrophenyl ethan-1-one hydrochloride,acmc-20amzi,2-amine-3'-nitroacetophenone hcl,2-amino-1-3-nitrophenyl ethanone hcl salt,2-amino-3-nitroacetophenone hydrochloride PubChem CID: 2774374 IUPAC Name: 2-amino-1-(3-nitrophenyl)ethanone;hydrochloride SMILES: [NH3+]CC(=O)C1=CC=CC(=C1)[N+]([O-])=O