missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117520
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15695G 5 g
A156925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15695G Supplier TCI America Supplier No. A15695G
Only null left

Chemical Identifiers

CAS 4389-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
InChI Key WNAJXPYVTFYEST-UHFFFAOYSA-N
Synonym 3-methylanthranilic acid, 2-amino-3-methyl-benzoic acid, 2-amino-3-methylbenzoicacid, 3-methyl-2-aminobenzoic acid, benzoic acid, 2-amino-3-methyl, 2-amino-m-toluic acid, 2-amino-3-methyl benzoic acid, m-toluic acid, 2-amino, unii-8rgx6sqe9n, 2-amino-3-methylbenzoate
PubChem CID 78101
ChEBI CHEBI:80574
IUPAC Name 2-amino-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1N)C(=O)O

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-methylbenzoic Acid