missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-methyl-5-nitropyridine 98.0+%, TCI America™
$46.60 - $139.81
Chemical Identifiers
| CAS | 18344-51-9 |
|---|---|
| Molecular Formula | C6H7N3O2 |
| Molecular Weight (g/mol) | 153.141 |
| InChI Key | JLPYUDJJBGYXEZ-UHFFFAOYSA-N |
| Synonym | 2-amino-3-methyl-5-nitropyridine, 2-amino-5-nitro-3-picoline, 3-methyl-5-nitro-pyridin-2-ylamine, 3-methyl-5-nitro-2-pyridylamine, 2-pyridinamine,3-methyl-5-nitro, 3-methyl-5-nitro-2-pyridinamine, 3-methyl-5-nitro-pyridine-2-ylamine, 3-methyl-5-nitro-pyridin-2-amine, 2-amino-5-nitro-3-picoline 2-amino-3-methyl-5-nitropyridine, pubchem1274 |
| PubChem CID | 2734446 |
| IUPAC Name | 3-methyl-5-nitropyridin-2-amine |
| SMILES | CC1=CC(=CN=C1N)[N+](=O)[O-] |
Chemical Identifiers
| 18344-51-9 | |
| 153.141 | |
| 2-amino-3-methyl-5-nitropyridine, 2-amino-5-nitro-3-picoline, 3-methyl-5-nitro-pyridin-2-ylamine, 3-methyl-5-nitro-2-pyridylamine, 2-pyridinamine,3-methyl-5-nitro, 3-methyl-5-nitro-2-pyridinamine, 3-methyl-5-nitro-pyridine-2-ylamine, 3-methyl-5-nitro-pyridin-2-amine, 2-amino-5-nitro-3-picoline 2-amino-3-methyl-5-nitropyridine, pubchem1274 | |
| 3-methyl-5-nitropyridin-2-amine |
| C6H7N3O2 | |
| JLPYUDJJBGYXEZ-UHFFFAOYSA-N | |
| 2734446 | |
| CC1=CC(=CN=C1N)[N+](=O)[O-] |
Specifications
| 18344-51-9 | |
| 261°C | |
| 1 g | |
| JLPYUDJJBGYXEZ-UHFFFAOYSA-N | |
| 3-methyl-5-nitropyridin-2-amine | |
| 2734446 | |
| ≥98.0% (GC,T) | |
| 2-Amino-3-methyl-5-nitropyridine |
| 255°C | |
| C6H7N3O2 | |
| 2-amino-3-methyl-5-nitropyridine, 2-amino-5-nitro-3-picoline, 3-methyl-5-nitro-pyridin-2-ylamine, 3-methyl-5-nitro-2-pyridylamine, 2-pyridinamine,3-methyl-5-nitro, 3-methyl-5-nitro-2-pyridinamine, 3-methyl-5-nitro-pyridine-2-ylamine, 3-methyl-5-nitro-pyridin-2-amine, 2-amino-5-nitro-3-picoline 2-amino-3-methyl-5-nitropyridine, pubchem1274 | |
| CC1=CC(=CN=C1N)[N+](=O)[O-] | |
| 153.141 | |
| 153.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No