missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxyanthraquinone 98.0+%, TCI America™

Catalog No. A031525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A031525G 25 g
A0315500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A031525G Supplier TCI America Supplier No. A031525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117-77-1
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00059499
InChI Key CNWWMJSRHGXXAX-UHFFFAOYSA-N
PubChem CID 67029
IUPAC Name 2-amino-3-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C=C3C2=O)O)N

Specifications

Quantity 25 g
Formula Weight 239.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-hydroxyanthraquinone
EINECSNumber (4)-0717
TSCA No