accessibility menu, dialog, popup

UserName

H2NOOHO

CAS RN 117-77-1

IUPAC Name: 2-amino-3-hydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-amino-3-hydroxyanthracene-9,10-dione
Molecular Weight (g/mol): 239.23
Molecular Formula: C14H9NO3
InChi Key: CNWWMJSRHGXXAX-UHFFFAOYSA-N
SMILES: NC1=CC2=C(C=C1O)C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results