missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-cyanopyridine 98.0+%, TCI America™
$173.37 - $592.76
Chemical Identifiers
| CAS | 24517-64-4 |
|---|---|
| Molecular Formula | C6H6N3 |
| Molecular Weight (g/mol) | 120.13 |
| MDL Number | MFCD00128867 |
| InChI Key | YYXDQRRDNPRJFL-UHFFFAOYSA-O |
| Synonym | 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506 |
| PubChem CID | 818259 |
| IUPAC Name | 2-amino-3-cyanopyridin-1-ium |
| SMILES | NC1=[NH+]C=CC=C1C#N |
Chemical Identifiers
| 24517-64-4 | |
| 120.13 | |
| YYXDQRRDNPRJFL-UHFFFAOYSA-O | |
| 818259 | |
| NC1=[NH+]C=CC=C1C#N |
| C6H6N3 | |
| MFCD00128867 | |
| 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506 | |
| 2-amino-3-cyanopyridin-1-ium |
Specifications
| 24517-64-4 | |
| White-Yellow | |
| MFCD00128867 | |
| 3439 | |
| YYXDQRRDNPRJFL-UHFFFAOYSA-O | |
| 2-amino-3-cyanopyridin-1-ium | |
| 818259 | |
| ≥98.0% (GC,T) | |
| 2-Amino-3-cyanopyridine |
| 135°C | |
| C6H6N3 | |
| 5 g | |
| 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506 | |
| NC1=[NH+]C=CC=C1C#N | |
| 120.13 | |
| 119.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No