missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyano-5-iodopyridine, 97%

CAS: 1347815-41-1 | C6H4IN3 | 245.023 g/mol

$156.95 - $524.30

Chemical Identifiers

CAS 1347815-41-1
Molecular Formula C6H4IN3
Molecular Weight (g/mol) 245.023
MDL Number MFCD20040120
InChI Key NJLCSLOABAUGHW-UHFFFAOYSA-N
Synonym 2-amino-5-iodonicotinonitrile, 2-amino-3-cyano-5-iodopyridine, 2-amino-5-iodo-nicotinonitrile
PubChem CID 65815340
IUPAC Name 2-amino-5-iodopyridine-3-carbonitrile
SMILES C1=C(C(=NC=C1I)N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3419103
View Documents
Thermo Scientific Chemicals
H3419103
1 g
Each for $156.95
Only null left
 
AAH3419106
View Documents
Thermo Scientific Chemicals
H3419106
5 g
Each for $524.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1347815-41-1
245.023
NJLCSLOABAUGHW-UHFFFAOYSA-N
65815340
C1=C(C(=NC=C1I)N)C#N
C6H4IN3
MFCD20040120
2-amino-5-iodonicotinonitrile, 2-amino-3-cyano-5-iodopyridine, 2-amino-5-iodo-nicotinonitrile
2-amino-5-iodopyridine-3-carbonitrile

Specifications

1347815-41-1
C6H4IN3
1 g
2-amino-5-iodonicotinonitrile, 2-amino-3-cyano-5-iodopyridine, 2-amino-5-iodo-nicotinonitrile
C1=C(C(=NC=C1I)N)C#N
245.023
245.02
2-Amino-3-cyano-5-iodopyridine
193°C to 197°C
MFCD20040120
UN3439
NJLCSLOABAUGHW-UHFFFAOYSA-N
2-amino-5-iodopyridine-3-carbonitrile
65815340
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only