CAS RN 1347815-41-1
IUPAC Name:
2-amino-5-iodopyridine-3-carbonitrile
Synonyms:
2-amino-5-iodopyridine-3-carbonitrile
Molecular Weight (g/mol):
245.02
Molecular Formula:
C6H4IN3
InChi Key:
NJLCSLOABAUGHW-UHFFFAOYSA-N
SMILES:
NC1=NC=C(I)C=C1C#N
1
–
1
of
1
results
1
Promotions Available
2-Amino-3-cyano-5-iodopyridine, 97%
2-Amino-3-cyano-5-iodopyridine, 97%
CAS: 1347815-41-1 Molecular Formula: C6H4IN3 Molecular Weight (g/mol): 245.023 MDL Number: MFCD20040120 InChI Key: NJLCSLOABAUGHW-UHFFFAOYSA-N Synonym: 2-amino-5-iodonicotinonitrile,2-amino-3-cyano-5-iodopyridine,2-amino-5-iodo-nicotinonitrile PubChem CID: 65815340 IUPAC Name: 2-amino-5-iodopyridine-3-carbonitrile SMILES: C1=C(C(=NC=C1I)N)C#N