missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyano-5-iodopyridine, 97%

Catalog No. AAH3419103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3419103 1 g
AAH3419106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3419103 Supplier Thermo Scientific Chemicals Supplier No. H3419103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1347815-41-1 | C6H4IN3 | 245.023 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1347815-41-1
Molecular Formula C6H4IN3
Molecular Weight (g/mol) 245.023
MDL Number MFCD20040120
InChI Key NJLCSLOABAUGHW-UHFFFAOYSA-N
Synonym 2-amino-5-iodonicotinonitrile, 2-amino-3-cyano-5-iodopyridine, 2-amino-5-iodo-nicotinonitrile
PubChem CID 65815340
IUPAC Name 2-amino-5-iodopyridine-3-carbonitrile
SMILES C1=C(C(=NC=C1I)N)C#N

Specifications

Melting Point 193°C to 197°C
Quantity 1 g
UN Number UN3439
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 2-Amino-3-cyano-5-iodopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only