missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloropyrazine 98.0+%, TCI America™

Catalog No. A21431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21431G 1 g
A21435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21431G Supplier TCI America Supplier No. A21431G

Chemical Identifiers

CAS 6863-73-6
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD04114305
InChI Key AEVSSZHXGJAPIE-UHFFFAOYSA-N
Synonym 2-amino-3-chloropyrazine, 3-chloro-2-pyrazinamine, 3-chloro-pyrazin-2-ylamine, pyrazinamine, 3-chloro, 3-amino-pyrazinechloride, 3-amino-2-chloropyrazine, 3-chloropyrazin-2-ylamine, 3-amino-2-chloro-pyrazine, 3-chloropyrazine-2-ylamine, 2-chloro-3-aminepyrazine
PubChem CID 276224
IUPAC Name 3-chloropyrazin-2-amine
SMILES NC1=NC=CN=C1Cl

Specifications

Melting Point 168°C
Quantity 1 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chloropyrazine