accessibility menu, dialog, popup

UserName

CAS RN 6863-73-6

IUPAC Name: 3-chloropyrazin-2-amine
Synonyms: 3-chloropyrazin-2-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: AEVSSZHXGJAPIE-UHFFFAOYSA-N
SMILES: NC1=NC=CN=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results